Learn More
Analytical Sales & Services Canary-Safe Waste Cap, B83, with 4 Ports for Barbed Adapters
Catalog No. 5000807094
Encompass
Canary-Safe Waste Cap B83 for Nalgene Bottles 4 Ports for Barbed Adapter 1 Port for Charcoal Exhaust Filter
Catalog No. 50-008-07094 Supplier Analytical Sales & Services Supplier No. CW83004
Canary-Safe Waste Cap B83 for Nalgene Bottles 4 Ports for Barbed Adapter 1 Port for Charcoal Exhaust Filter