Learn More
Analytical Sales & Services Canary-Safe Waste Cap, B83, with Safety Funnel with Shut-Off Valve & 4 Standard Ports
Catalog No. 5000805274
Encompass
Canary-Safe Waste Cap B83 with Safety Funnel with Shut-Off Valve 4 Standard Ports 1 Exhaust Filter Port
Catalog No. 50-008-05274 Supplier Analytical Sales & Services Supplier No. CSF83418
Canary-Safe Waste Cap B83 with Safety Funnel with Shut-Off Valve 4 Standard Ports 1 Exhaust Filter Port