Learn More
Analytical Sales & Services Canary-Safe Waste Cap, GL45, with 1 Standard Tubing Port
Catalog No. 5000805358
Encompass
Canary-Safe Waste Cap GL45 1 Standard Tubing Port 1 Port for Charcoal Exhaust Filter
Catalog No. 50-008-05358 Supplier Analytical Sales & Services Supplier No. CW45018
Canary-Safe Waste Cap GL45 1 Standard Tubing Port 1 Port for Charcoal Exhaust Filter