Learn More
Analytical Sales & Services Canary-Safe Waste Cap, GL45, with 2 Ports for Barbed Adapter
Catalog No. 5000805262
Encompass
Canary-Safe Waste Cap GL45 with 2 Ports for Barbed Adapter 1 Port for Charcoal Exhaust Filter
Catalog No. 50-008-05262 Supplier Analytical Sales & Services Supplier No. CW45002
Canary-Safe Waste Cap GL45 with 2 Ports for Barbed Adapter 1 Port for Charcoal Exhaust Filter