Learn More
Analytical Sales & Services Canary-Safe Waste Cap GL45, 2 Standard Tubing Ports, 2 Ports for Barbed Adapters
Catalog No. 5000805263
Encompass
Canary-Safe Waste Cap GL45, 2 Standard Tubing Ports, 2 Ports for Barbed Adapters, 1 Port for Charcoal Exhaust Filter
Catalog No. 50-008-05263 Supplier Analytical Sales & Services Supplier No. CW452182
Canary-Safe Waste Cap GL45, 2 Standard Tubing Ports, 2 Ports for Barbed Adapters, 1 Port for Charcoal Exhaust Filter