Learn More
Analytical Sales & Services Canary-Safe Waste Cap, GL45, with 3 Standard Tubing Ports ONLY
Catalog No. 5000807144
Encompass
Canary-Safe Waste Cap GL45 3 Standard Tubing Ports ONLY (no port for exhaust filter).
Catalog No. 50-008-07144 Supplier Analytical Sales & Services Supplier No. CW45313
Add to Cart
Add to Cart
Canary-Safe Waste Cap GL45 3 Standard Tubing Ports ONLY (no port for exhaust filter).