Learn More
Analytical Sales & Services Canary-Safe Waste Cap, GL45, with 3 Standard Tubing Ports
Catalog No. 5000805276
Encompass
Canary-Safe Waste Cap GL45 3 Standard Tubing Ports 1 Port for Charcoal Exhaust Filter
Catalog No. 50-008-05276 Supplier Analytical Sales & Services Supplier No. CW45319
Canary-Safe Waste Cap GL45 3 Standard Tubing Ports 1 Port for Charcoal Exhaust Filter