Learn More
Analytical Sales & Services Canary-Safe Waste Cap, S55, with 2 Standard Tubing Ports & 1 Port for Barbed Adapter
Catalog No. 5000805359
Encompass
Canary-Safe Waste Cap S55 2 Standard Tubing Ports 1 Port for Barbed Adapter 1 Port for Charcoal Exhaust Filter
Catalog No. 50-008-05359 Supplier Analytical Sales & Services Supplier No. CW55218
Canary-Safe Waste Cap S55 2 Standard Tubing Ports 1 Port for Barbed Adapter 1 Port for Charcoal Exhaust Filter