Learn More
Analytical Sales & Services Canary-Safe Waste Cap, S60, with 2 Standard Tubing Ports & 1 Port for Barbed Adapter
Catalog No. 5000805360
Encompass
Canary-Safe Waste Cap S60 2 Standard Tubing Ports 1 Port for Barbed Adapter 1 Port for Charcoal Exhaust Filter
Catalog No. 50-008-05360 Supplier Analytical Sales & Services Supplier No. CW60218
Canary-Safe Waste Cap S60 2 Standard Tubing Ports 1 Port for Barbed Adapter 1 Port for Charcoal Exhaust Filter