Learn More
Analytical Sales & Services Canary-Safe Waste Cap, S90, with 4 Standard Tubing Ports & 1 Port for Barbed Adapter
Catalog No. 5000805372
Encompass
Canary-Safe Waste Cap S90 4 Standard Tubing Ports 1.Port for Barbed Adapter 1 Port for Charcoal Exhaust Filter
Catalog No. 50-008-05372 Supplier Analytical Sales & Services Supplier No. CW90418
Canary-Safe Waste Cap S90 4 Standard Tubing Ports 1.Port for Barbed Adapter 1 Port for Charcoal Exhaust Filter