Learn More
Enzo Life Sciences Canavanine . sulfate (500mg). CAS: 2219-31-0
SDP
Catalog No. 502008678
Encompass_Preferred
The non-protein amino acid L-canavanine is an analog of L-arginine. Selective inducible nitric oxide synthase (iNOS; NOS II) inhibitor. Induces apoptotic cell death and shows antiproliferative and immunotoxic effects. Alternative name: L-alpha-Amino-gamma-(guanidinooxy)-n-butyric acid . sulfate. Purity: ≥99% (TLC). Appearance: White to-off white powder. Solubility: Soluble in water (100mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-8678 Supplier Enzo Life Sciences Supplier No. ALX350002M500
Add to Cart
Add to Cart
The non-protein amino acid L-canavanine is an analog of L-arginine. Selective inducible nitric oxide synthase (iNOS; NOS II) inhibitor. Induces apoptotic cell death and shows antiproliferative and immunotoxic effects. Alternative name: L-alpha-Amino-gamma-(guanidinooxy)-n-butyric acid . sulfate. Purity: ≥99% (TLC). Appearance: White to-off white powder. Solubility: Soluble in water (100mg/ml). Long Term Storage: +4°C.