Learn More
Med Vet International Cancellous Lag Screw 4.0mm x 22mm L, J0425G, Each
Catalog No. 5000582958
Encompass_Preferred
4mm Cancellous (Part Threaded) Lag Screw. For use as a lag screw without overdrilling of proximal fragment. 1/pkg.
Catalog No. 50-005-82958 Supplier Med Vet International Supplier No. JOR425G
4mm Cancellous (Part Threaded) Lag Screw. For use as a lag screw without overdrilling of proximal fragment. 1/pkg.