Learn More
Med Vet International Cancellous Lag Screw 4.0mm x 26mm L, J0425I, Each
Catalog No. 5000582561
Encompass_Preferred
4mm Cancellous (Part Threaded) Lag Screw. For use as a lag screw without overdrilling of proximal fragment. 1/pkg.
Catalog No. 50-005-82561 Supplier Med Vet International Supplier No. JOR425I
4mm Cancellous (Part Threaded) Lag Screw. For use as a lag screw without overdrilling of proximal fragment. 1/pkg.