Learn More
Med Vet International Cancellous Lag Screw 4.0mm x 28mm L, J0425J, Each
Catalog No. 5000582383
Encompass_Preferred
4mm Cancellous (Part Threaded) Lag Screw. For use as a lag screw without overdrilling of proximal fragment. 1/pkg.
Catalog No. 50-005-82383 Supplier Med Vet International Supplier No. JOR425J
4mm Cancellous (Part Threaded) Lag Screw. For use as a lag screw without overdrilling of proximal fragment. 1/pkg.