Learn More
Med Vet International Cancellous Lag Screw 4.0mm x 40mm L, J0425M, Each
Catalog No. 5000582754
Encompass_Preferred
4mm Cancellous (Part Threaded) Lag Screw. For use as a lag screw without overdrilling of proximal fragment. 1/pkg.
Catalog No. 50-005-82754 Supplier Med Vet International Supplier No. JOR425M
4mm Cancellous (Part Threaded) Lag Screw. For use as a lag screw without overdrilling of proximal fragment. 1/pkg.