Learn More
Med Vet International Cancellous Screw, J0419MC, 3mm x 30mm, Each
Catalog No. 5000582846
Encompass_Preferred
2mm pilot. 2.5 hex head. Self-tapping. Single pack.
Catalog No. 50-005-82846 Supplier Med Vet International Supplier No. JOR419MC
2mm pilot. 2.5 hex head. Self-tapping. Single pack.