Learn More
Med Vet International Cancellous Screw, J0419NC, 3mm x 32mm, Each
Catalog No. 5000582706
Encompass_Preferred
2mm pilot. 2.5 hex head. Self-tapping. Single pack.
Catalog No. 50-005-82706 Supplier Med Vet International Supplier No. JOR419NC
Add to Cart
Add to Cart
2mm pilot. 2.5 hex head. Self-tapping. Single pack.