Learn More
Med Vet International Cancellous Screw, J0419OC, 3mm x 34mm, Each
Catalog No. 5000582923
Encompass_Preferred
2mm pilot. 2.5 hex head. Self-tapping. Single pack.
Catalog No. 50-005-82923 Supplier Med Vet International Supplier No. JOR419OC
2mm pilot. 2.5 hex head. Self-tapping. Single pack.