Learn More
Med Vet International Cancellous Screws 3.5mm x 20mm, J0423F, 6 Pack
Catalog No. 5000582646
Encompass_Preferred
3.5mm Cancellous Screw. 1.75 Pitch. The coarser thread has a much bigger "bite" into the bone. Use on metaphyseal or other cancellous bone. The screw needs its own tap. (JOR409c)
Catalog No. 50-005-82646 Supplier Med Vet International Supplier No. JOR423F
3.5mm Cancellous Screw. 1.75 Pitch. The coarser thread has a much bigger "bite" into the bone. Use on metaphyseal or other cancellous bone. The screw needs its own tap. (JOR409c)