Learn More
Biochain Institute Inc CancerSeq Paraffin Tissue Tumor Block: Colon, 1 Block/pk
SDP
Catalog No. 501871558
Encompass_Preferred
To generate our CancerSeq line of tissues, genomic DNA was extracted from the FFPE tissues and validated by targeted NGS with Illumina's TruSeq Amplicon - Cancer Panel (TSACP), which includes 48 cancer-related genes. Our prescreened tissues come with detailed information regarding donor and cancer type, as well as NGS information such as chromosomal position, variant type (SNP or indel), alternate allele variations, quality score, depth of coverage, allele type, transcript ID and more. We offer the prescreened tissue as mounted sections or whole block. Please inquire for other options, such as curls or extracted nucleic acids.
Catalog No. 50-187-1558 Supplier Biochain Institute Inc Supplier No. T2235090SB
To generate our CancerSeq line of tissues, genomic DNA was extracted from the FFPE tissues and validated by targeted NGS with Illumina's TruSeq Amplicon - Cancer Panel (TSACP), which includes 48 cancer-related genes. Our prescreened tissues come with detailed information regarding donor and cancer type, as well as NGS information such as chromosomal position, variant type (SNP or indel), alternate allele variations, quality score, depth of coverage, allele type, transcript ID and more. We offer the prescreened tissue as mounted sections or whole block. Please inquire for other options, such as curls or extracted nucleic acids.