Learn More
Aviva Systems Biology Corp CAND1 Antibody - middle region (ARP34584_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34584_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAND1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CAND1. Alternate names: DKFZp434M1414, FLJ10114, FLJ10929, FLJ38691, FLJ90441, KIAA0829, TIP120, TIP120AApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55462