Learn More
ATCC Candida albicans strain 3147 (ATCC 10231™)
Catalog No. 502382046
Encompass
Candida albicans strain 3147 (ATCC 10231™) is a reference yeast strain used in sterility, preservative efficacy, and antifungal testing. This strain is recommended as a test organisms in USP <51>, <61>, <62>, <71>, and <2021>.
Recommended companion products:
MicroQuant™Candida albicans, high CFU (Fisher Cat. No. NC3456076, ATCC 10231-HQ-PACK™),
MicroQuant™ Candida albicans, low CFU (Fisher Cat. No. NC3456089, ATCC 10231-LQ-PACK™)
Genomic DNA (Fisher Cat. No. 50-238-4704, ATCC 10231DQ™).
Catalog No. 50-238-2046 Supplier ATCC Supplier No. 10231
 Restricted Item
Add to Cart
Add to Cart
Candida albicans strain 3147 (ATCC 10231™) is a reference yeast strain used in sterility, preservative efficacy, and antifungal testing. This strain is recommended as a test organisms in USP <51>, <61>, <62>, <71>, and <2021>.
Recommended companion products:
MicroQuant™Candida albicans, high CFU (Fisher Cat. No. NC3456076, ATCC 10231-HQ-PACK™),
MicroQuant™ Candida albicans, low CFU (Fisher Cat. No. NC3456089, ATCC 10231-LQ-PACK™)
Genomic DNA (Fisher Cat. No. 50-238-4704, ATCC 10231DQ™).