Learn More
ATCC Candida albicans; genomic DNA
Catalog No. 502382183
Encompass
Genomic DNA isolated from Candida albicans strain 3147 (ATCC 10231). This whole-genome sequenced product can be used in assay development, PCR-based assays, and other molecular biology applications
Catalog No. 50-238-2183 Supplier ATCC Supplier No. 10231D5
Genomic DNA isolated from Candida albicans strain 3147 (ATCC 10231). This whole-genome sequenced product can be used in assay development, PCR-based assays, and other molecular biology applications