Learn More
Drg International Inc DRG Candida albicans IgA ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3457R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgA-class antibodies to Candida albicans in serum. This product is for Research Use Only. The Candida albicans IgA ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA). Microtiter wells as a solid phase are coated with Candida albicans antigen. Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-187-1243