Learn More
ATCC Candida albicans; Quantitative DNA
Catalog No. 502384704
Encompass
Quantitative genomic DNA from Candida albicans strain 3147 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load and for other molecular biology applications.
Catalog No. 50-238-4704 Supplier ATCC Supplier No. 10231DQ
Quantitative genomic DNA from Candida albicans strain 3147 can be used for assay development, verification, and validation as well as monitoring of day-to-day test variation and lot-to-lot performance of molecular-based assays. The quantitative format allows for the generation of a standard curve for quantitative PCR (qPCR) to determine fungal load and for other molecular biology applications.