Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Candida krusei, Genomic DNA
SDP

Catalog No. 21100512 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-100-512 1 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-100-512 Supplier ZeptoMetrix Supplier No. 0801536DNA1UG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use as an amplification/detection control for nucleic acid testing of Candida krusei

  • Available in 1μg aliquot
  • Supplied in TE buffer

Determine limit of detection (LOD) in diagnostic assay development, cross-reactivity studies or genomic sequencing

Specifications

Certifications/Compliance ISO, 13485
Content And Storage -20°C
Media Type Vial
Product Type Genomic DNA Bacteria Control
For Use With (Application) Cross-reactivity studies, limit of detection studies, Nucleic Acid or molecular testing, other viral based assays
Quantity 1 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.