Learn More
HOLLISTER STIER LABORATORIES LLC CANDIDA SCRATCH TEST
Catalog No. NC9009112
Encompass
Allergenic extract Candida albicans. 5mL scratch diagnostic 50% Glycerin. 1-10 W/V. Requires refrigeration at 2 - 8 degrees celcius.
Catalog No. NC9009112 Supplier HOLLISTER STIER LABORATORIES LLC Supplier No. 5053ED
Allergenic extract Candida albicans. 5mL scratch diagnostic 50% Glycerin. 1-10 W/V. Requires refrigeration at 2 - 8 degrees celcius.