Learn More
Carbosynth LLC. Candidalysin (0.5 mg vial), 1ea, MW: 3310.1, Molecular formula: C153H266N38O38S2, CAS# 1906866-53-2
SDP
Supplier:  Carbosynth LLC. PCT4494V0.5MG
Encompass_Preferred
Synonyms: Ecel-III62-92K, Ser-Ile-Ile-Gly-Ile-Ile-Met-Gly-Ile-Leu-Gly-Asn-Ile-Pro-Gln-Val-Ile-Gln-Ile-Ile-Met-Ser-Ile-Val-Lys-Ala-Phe-Lys-Gly-Asn-Lys, Purity: , ≥95.0% (HPLC), Application: Fungal Cytolytic Peptide Toxin, Salt Form:, Trifluoroacetate, Storage Temperature: -20 °C, References: D.L. Moyes, et al., Nature, 532, 64 (2016). (Original), A.P. Mitchell, Nature, 532, 41 (2016). (News & Views), U. Hofer, Nature Rev. Microbiol., 14, 268 (2016). (Research Highlights)
Catalog No. 50-168-6411
 Restricted Item