Learn More
Boca Scientific Inc Candor ELISA Sampler Package 2 COVID-19
Supplier:  Boca Scientific Inc 600 031
Encompass
Sensitivity and specificity of antibody detection assays for COVID-19 continue to present challenges. This sampler package contains all the reagents you need for reliable serological ELISA, complete with blockers, stabilizers and optimizers. Six reagents are included: 125ml of each reagent is included: PlateBlock™, a blocker specifically designed for serological ELISA, LowCross-Buffer STRONG, an innovative assay buffer that reduces interference, Liquid Plate Sealer Animal-Free, a stabilizer for coated surfaces, HRP-Protector™, a stabilizer for HRP-conjugates, and Coating Buffer (pH 7.4), a buffer for adsorptive immobilization of proteins and antibodies on plastic surfaces. These reagents will greatly improve the accuracy of your assay and significantly reduce false positives and false negatives.
Catalog No. NC1894481