Learn More
Medchemexpress LLC HY-10367 50mg Medchemexpress, Canertinib CAS:267243-28-7 Purity:>98%
SDP
Catalog No. 501968159
Encompass_Preferred
Medchemexpress, HY-10367 50mg Canertinib CAS:267243-28-7 Canertinib (CI-1033;PD-183805) is a potent and irreversible EGFR inhibitor; inhibits cellular EGFR and ErbB2 autophosphorylation with IC50s of 7.4 and 9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8159 Supplier Medchemexpress LLC Supplier No. HY1036750MG
Medchemexpress, HY-10367 50mg Canertinib CAS:267243-28-7 Canertinib (CI-1033;PD-183805) is a potent and irreversible EGFR inhibitor; inhibits cellular EGFR and ErbB2 autophosphorylation with IC50s of 7.4 and 9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.