Learn More
Abcam Canertinib (CI-1033), EGFR and HER2/ErbB2 tyrosine kinase inhibitor, 50MG
Catalog No. 502706650
Encompass_Preferred
MW 558.9 Da, Purity >99%. Potent, irreversible pan-ErbB tyrosine kinase inhibitor (IC₅₀ values are 1.5 and 9 nM at EGFR and ErbB2 respectively). Orally active. Antitumor and radiation-sensitizing agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6650 Supplier Abcam Supplier No. AB14140250MG
MW 558.9 Da, Purity >99%. Potent, irreversible pan-ErbB tyrosine kinase inhibitor (IC₅₀ values are 1.5 and 9 nM at EGFR and ErbB2 respectively). Orally active. Antitumor and radiation-sensitizing agent. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement