Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Canertinib dihydrochloride | 289499-45-2 | MFCD09954112 | 558.86 g/mol | C24H27Cl3FN5O3 | 500 MG
SDP

Catalog No. 5000245209
Encompass_Preferred

Canertinib dihydrochloride is an irreversible inhibitor of epidermal growth factor receptor (EGFR) and ErbB2 (HER2) tyrosine kinases used in preclinical biochemical and cellular studies to probe receptor signaling and kinase inhibition mechanisms.

  • Irreversible EGFR and ErbB2 inhibition suitable for kinase studies.
  • Applicable to biochemical and cellular assay workflows.
  • Molecular formula C24H27Cl3FN5O3; molecular weight 558.86 g/mol.
  • Storage: keep sealed and away from moisture; in solution, store frozen as recommended.
  • Provided as a research-grade reagent; consult the product datasheet for handling and safety information.

Catalog No. 50-002-45209 Supplier Medchemexpress LLC Supplier No. HY10367A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Canertinib dihydrochloride is an irreversible inhibitor of epidermal growth factor receptor (EGFR) and ErbB2 (HER2) tyrosine kinases used in preclinical biochemical and cellular studies to probe receptor signaling and kinase inhibition mechanisms.

  • Irreversible EGFR and ErbB2 inhibition suitable for kinase studies.
  • Applicable to biochemical and cellular assay workflows.
  • Molecular formula C24H27Cl3FN5O3; molecular weight 558.86 g/mol.
  • Storage: keep sealed and away from moisture; in solution, store frozen as recommended.
  • Provided as a research-grade reagent; consult the product datasheet for handling and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.