Learn More
TOYOBO USA INC Can Get Signal | 250mL | Solution 1
Catalog No. NC1859569
Encompass
Can Get Signal is a solution containing an accelerator for antigen-antibody reactions. This reagent improves sensitivity, specificity, and signal-to-noise ration (S/N) for Western blotting, dot blotting, enzyme-linked immunosorbent assay (ELISA), etc. Solutions 1 and 2 refer to the reactions of the primary and secondary antibodies, respectively.
Catalog No. NC1859569 Supplier TOYOBO USA INC Supplier No. NKB201
Can Get Signal is a solution containing an accelerator for antigen-antibody reactions. This reagent improves sensitivity, specificity, and signal-to-noise ration (S/N) for Western blotting, dot blotting, enzyme-linked immunosorbent assay (ELISA), etc. Solutions 1 and 2 refer to the reactions of the primary and secondary antibodies, respectively.