Learn More
Selleck Chemical LLC Cangrelor Tetrasodium 5mg 163706-36-3 AR-C69931MX
SDP
Supplier:  Selleck Chemical LLC S37375MG
Encompass_Preferred
Cangrelor (AR-C69931MX) is a potent, competitive P2Y12 receptor inhibitor that is administered by intravenous infusion and rapidly achieves near complete inhibition of ADP-induced platelet aggregation. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8278