Learn More
ATCC Canid herpesvirus 1; Canine herpesvirus; Strain: D 004
Supplier:  ATCC VR552
Encompass
Canid herpesvirus 1 (canine herpesvirus) strain D 004 is propagated in MDCK (NBL-2) cells (ATCC CCL-34). This strain was isolated from the respiratory tract of a dog and was deposited by the Division of Veterinary Medicine at Walter Reed Army Institute of Research.
Catalog No. 50-238-4831