Learn More
ACROBiosystems ACROBiosystems Canine 4-1BB / TNFRSF9 Protein, Fc Tag (MALS verified)
Catalog No. 502106425
Encompass_Preferred
Recombinant Protein;100UG;Canine 4-1BB Protein, Fc Tag (41B-C5253) is expressed from human 293 cells (HEK293). It contains AA Ile 24 - Ser 185 (Accession # XP_850336.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 43.4 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 4-1BB Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. 41L-H5254) at 5 μg/mL (100 μL/well) can bind Canine 4-1BB Protein, Fc Tag (Cat. No. 41B-C5253) with a linear range of 0.3-5 ng/mL (QC tested).
Catalog No. 50-210-6425 Supplier ACROBiosystems Supplier No. 41BC5253100UG
Recombinant Protein;100UG;Canine 4-1BB Protein, Fc Tag (41B-C5253) is expressed from human 293 cells (HEK293). It contains AA Ile 24 - Ser 185 (Accession # XP_850336.1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 43.4 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human 4-1BB Ligand, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. 41L-H5254) at 5 μg/mL (100 μL/well) can bind Canine 4-1BB Protein, Fc Tag (Cat. No. 41B-C5253) with a linear range of 0.3-5 ng/mL (QC tested).