Learn More
American Research Products Inc Canine acetylcholine receptor
Catalog No. 5000329709
Encompass
Canine acetylcholine receptor antibody AChRab ELISA Kit from CUSABIO Detection Range 3 75 ng/mL-240 ng/mL Sensitivity 0 94 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-29709 Supplier American Research Products Inc Supplier No. CSBE08375C
Canine acetylcholine receptor antibody AChRab ELISA Kit from CUSABIO Detection Range 3 75 ng/mL-240 ng/mL Sensitivity 0 94 ng/mL Sample serum plasma Method Sandwich