Learn More
Raybiotech Inc Canine Albumin ELISA Kit
Supplier:  Raybiotech Inc ELCALBUMIN1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 14 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Canine Albumin.
Catalog No. 50-194-8876