Learn More
Kingfisher Biotech Inc Canine Amphiregulin Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP1846D005
Encompass
The Canine Amphiregulin yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine Amphiregulin applications are for cell culture, ELISA standard, and Western Blot Control. Canine Amphiregulin yeast-derived recombinant protein can be purchased in multiple sizes. Canine Amphiregulin Specifications: (Molecular Weight: 10.1 kDa) (Amino Acid Sequence: SVRVEQVVKP MKNKTESEKT SDKPKRKKKG GKSGKNRRNR KKKNPCDAEF QNFCIHGECK YIEHLEAVTC QCHQDYFGER CGEKSMK (87)) (Gene ID: 475176). For research use only. Made in the USA
Catalog No. 50-253-2700