Learn More
Kingfisher Biotech Inc Canine BAFF (TNFSF13B) Recombinant Protein (Yeast, Endotoxin - Free) - 25 ug
SDP
Supplier:  Kingfisher Biotech Inc RP0465D025
Encompass
The Canine BAFF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine BAFF applications are for cell culture, ELISA standard, and Western Blot Control. The Canine BAFF yeast-derived recombinant protein can be purchased in multiple sizes. Canine BAFF Specifications: (Molecular Weight: 17.1 kDa) (Amino Acid Sequence: AIQGPEETVT QDCLQLIADS DTPTIRKGAY TFVPWLLSFK RGRALEEKEN KILVKEAGYF FIYSQVLYTD NTFAMGHLIQ RKKVHVFGDE LSLVTLFRCI QNMPETLPNN SCYSAGIAKL EEGDELQLAI PREDAKISRD GDGTFFGALK LL (152)) (Gene ID: 485545). For research use only. Made in the USA
Catalog No. 50-241-3332