Learn More
Innovative Research Inc Canine Beagle IgG Affinity Purified, 50mg
Catalog No. NC3316642
Encompass
Canine Beagle IgG Affinity Purified from Innovative Research has been purified from normal Beagle Serum using Protein A affinity chromatography. This is a frozen liquid buffered in 0.02M Sodium Phosphate; 0.15M NaCl; pH 7.4 and a purity of >95%. This product is useful for ELISA, Western Blot, and as an isotype control. More Details: Source: Beagle Serum Purification: IgG Fractionated Purity: >95% Storage Conditions: -70 C At Innovative Research, we provide reliable, consistent products that deliver reliable, consistent results. Please note ¢ this product replaces our old product number IR-BG-GF (same product, new number).
Catalog No. NC3316642 Supplier Innovative Research Inc Supplier No. ICNBGIGGAP50MG
Canine Beagle IgG Affinity Purified from Innovative Research has been purified from normal Beagle Serum using Protein A affinity chromatography. This is a frozen liquid buffered in 0.02M Sodium Phosphate; 0.15M NaCl; pH 7.4 and a purity of >95%. This product is useful for ELISA, Western Blot, and as an isotype control. More Details: Source: Beagle Serum Purification: IgG Fractionated Purity: >95% Storage Conditions: -70 C At Innovative Research, we provide reliable, consistent products that deliver reliable, consistent results. Please note ¢ this product replaces our old product number IR-BG-GF (same product, new number).