Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creative Bioarray Canine Bone Marrow Neutrophils, derived from tibia of beagle dog, normal, healthy cells

Catalog No. NC2832811
Encompass

Creative Bioarray's Canine Bone Marrow Neutrophils are primary cells derived from the tibia of beagle dogs, making them essential for research in canine immunology, hematopoiesis, and inflammatory responses. These healthy neutrophils are carefully quality-controlled to guarantee they are free from common contaminants, ensuring reliable and reproducible experimental outcomes. They are shipped in frozen conditions for optimal preservation.

  • Species: Dog
  • Source: Bone marrow
  • Cell type: Neutrophil, granulocyte
  • Disease state: Normal
  • Derivation: Tibia of beagle dog
  • Recommended medium: SuperCult Complete Neutrophil Medium
  • Quality control: Negative for bacteria, yeast, fungi, mycoplasma
  • Shipping condition: Frozen on dry ice

Catalog No. NC2832811 Supplier Creative Bioarray Supplier No. CSCC4843L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Creative Bioarray's Canine Bone Marrow Neutrophils are primary cells derived from the tibia of beagle dogs, making them essential for research in canine immunology, hematopoiesis, and inflammatory responses. These healthy neutrophils are carefully quality-controlled to guarantee they are free from common contaminants, ensuring reliable and reproducible experimental outcomes. They are shipped in frozen conditions for optimal preservation.

  • Species: Dog
  • Source: Bone marrow
  • Cell type: Neutrophil, granulocyte
  • Disease state: Normal
  • Derivation: Tibia of beagle dog
  • Recommended medium: SuperCult Complete Neutrophil Medium
  • Quality control: Negative for bacteria, yeast, fungi, mycoplasma
  • Shipping condition: Frozen on dry ice

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.