Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Canine C-peptide ELISA

Catalog No. EZCCP47K Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
96 Wells/Strip Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EZCCP47K 96 Wells/Strip Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. EZCCP47K Supplier MilliporeSigma™ Supplier No. EZCCP47K
Add to Cart
Add to Cart

Measures, quantifies C-Peptide levels in metabolism , endocrine research

  • Detection method: Fluorescent, Colorimetric
  • Precision%: Inter Assay: <15%, Intra Assay:<10%
  • Reactivity: Canine
  • Analytes Available: C-Peptide
TRUSTED_SUSTAINABILITY

Specifications

Quantity 96 Wells/Strip Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.