Learn More
Abcam Canine C Reactive Protein ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1576981X96TESTS
Encompass_Preferred
Canine C Reactive Protein ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Canine C Reactive Protein in Dog in Plasma, Serum, Other biological fluids samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2394094