Learn More
American Research Products Inc Canine C ReactiVe Protein CRP
Catalog No. 5000329454
Encompass
Canine C-ReactiVe Protein CRP ELISA Kit from CUSABIO Detection Range 0 312 g/mL-20 g/mL Sensitivity 0 078 g/mL Sample serum plasma cell culture supernates Method Sandwich
Catalog No. 50-003-29454 Supplier American Research Products Inc Supplier No. CSBE14262C
Canine C-ReactiVe Protein CRP ELISA Kit from CUSABIO Detection Range 0 312 g/mL-20 g/mL Sensitivity 0 078 g/mL Sample serum plasma cell culture supernates Method Sandwich