Learn More
American Research Products Inc Canine calprotectin CALP ELI
Catalog No. 5000327952
Encompass
Canine calprotectin (CALP) ELISA kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27952 Supplier American Research Products Inc Supplier No. CSBEQ013485DO
Canine calprotectin (CALP) ELISA kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 1 56 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich