Learn More
American Research Products Inc Canine cardiac isoform of Trop
Catalog No. 5000326736
Encompass
Canine cardiac isoform of Tropnin T cTnT ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26736 Supplier American Research Products Inc Supplier No. CSBE14377C
Canine cardiac isoform of Tropnin T cTnT ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich