Learn More
American Research Products Inc Canine Cardiac Troponin I
Catalog No. 5000329207
Encompass
Canine Cardiac Troponin (cTn- ) ELISA kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29207 Supplier American Research Products Inc Supplier No. CSBE08772C
Canine Cardiac Troponin (cTn- ) ELISA kit from CUSABIO Detection Range 23 5 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates Method Sandwich