Learn More
Kingfisher Biotech Inc Canine CCL17 Recombinant Protein (Yeast, Endotoxin - Free) - 5 ug
SDP
Supplier:  Kingfisher Biotech Inc RP2080D005
Encompass
The Canine CCL17 yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Canine CCL17 applications are for cell culture, ELISA standard, and Western Blot Control. Canine CCL17 yeast-derived recombinant protein can be purchased in multiple sizes. Canine CCL17 Specifications: (Molecular Weight: 8.6 kDa) (Amino Acid Sequence: ARGTNVGREC CLEYFKGAIP ISRLTRWYKT SGECPKDAIV FVTVQGKSIC SDPKDKRVKK AVRYLQRTWK GGPQES (76)) (Gene ID: 403586). For research use only. Made in the USA
Catalog No. 50-253-2467