Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems Canine CD3 Epsilon Protein, His Tag, 1 mg (MALS-verified)

Catalog No. NC3740095
Encompass_Preferred

Canine CD3 epsilon Protein is a recombinant extracellular domain of canine CD3ε (Gln 22-Leu 122) produced in HEK293 cells with a C-terminal polyhistidine tag. The material is MALS-verified and supplied as a 1 mg format (500 μg x 2) with high purity for biochemical characterization and binding studies.

  • Expressed in HEK293 (human 293) cells.
  • Contains amino acids Gln 22-Leu 122.
  • C-terminal polyhistidine tag present.
  • Purity >90% by SDS-PAGE and >95% by SEC-MALS.
  • Calculated molecular weight 16.7 kDa; migrates at 19-25 kDa by SDS-PAGE.
  • Verified by SEC-MALS.
  • Supplied as 1 mg (500 μg x 2) for research use.

Catalog No. NC3740095 Supplier ACROBiosystems Supplier No. CDEC52H31MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Canine CD3 epsilon Protein is a recombinant extracellular domain of canine CD3ε (Gln 22-Leu 122) produced in HEK293 cells with a C-terminal polyhistidine tag. The material is MALS-verified and supplied as a 1 mg format (500 μg x 2) with high purity for biochemical characterization and binding studies.

  • Expressed in HEK293 (human 293) cells.
  • Contains amino acids Gln 22-Leu 122.
  • C-terminal polyhistidine tag present.
  • Purity >90% by SDS-PAGE and >95% by SEC-MALS.
  • Calculated molecular weight 16.7 kDa; migrates at 19-25 kDa by SDS-PAGE.
  • Verified by SEC-MALS.
  • Supplied as 1 mg (500 μg x 2) for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.